Principaux documents
Z740710
Sigma-Aldrich
Aldrich® Essentials boiling flask, round-bottom, ST/NS 24/40
capacity 1000 mL
1 EA
51.60 CHF
51.60 CHF
Check Cart for Availability
Changer de vue
1 EA
51.60 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
51.60 CHF
Check Cart for Availability
Matériaux
borosilicate glass flask
round bottom
single-neck
Conditionnement
pkg of 1 ea
Capacité
1000 mL
Description générale
Clear borosilicate glass, round bottomed boiling flasks with outer ST ground joint neck. White marking spot.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC