Principaux documents
Z682853
Ace receiving flask,
capacity 500 mL, joint: ST/NS 35/20, plastic-coated glass
1 EA
108.00 CHF
108.00 CHF
Check Cart for Availability
Changer de vue
1 EA
108.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
108.00 CHF
Check Cart for Availability
Matériaux
plastic-coated glass
round bottom flask
single-neck
Joint
joint: ST/NS35/20
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Ace Glass 3996-08
Capacité
500 mL
Description générale
The receiving flasks have the standard joint to fit the glassware used in rotary evaporators, and are available with poly-coating or plain glass.