Principaux documents
Z682632
Ace evaporating flask, pear-shaped
capacity 2000 mL, joint: ST/NS 29/32, poly-coated, heavy wall
Synonyme(s) :
Aldrich® evaporating flask, pear-shaped, bottom flask, flask round, round flask
1 EA
128.00 CHF
128.00 CHF
Check Cart for Availability
Changer de vue
1 EA
128.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
128.00 CHF
Check Cart for Availability
Matériaux
borosilicate glass
pear-shaped flask
single-neck
Joint
joint: ST/NS29/32
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Büchi equivalent, 25323
Capacité
2000 mL
Catégories apparentées
Description générale
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC