Principaux documents
Z682195
Ace receiving flask adapter
joint: ST/NS 24/40, fits Büchi® models R-114/R-144
1 EA
220.00 CHF
220.00 CHF
Check Cart for Availability
Changer de vue
1 EA
220.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
220.00 CHF
Check Cart for Availability
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Büchi equivalent, 23747
Catégories apparentées
Description générale
The receiving flask adapter fits condenser assemblies "C", "CR", "A", and "V" for Büchi® evaporator models R-200/205. Adapter clip: not included.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG