Principaux documents
Z677396
Dermatril P nitrile gloves
size L
Synonyme(s) :
Dermatril P Gloves, Laboratory Nitrile Gloves, Nitrile Laboratory Gloves, disposable glove
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
blue gloves
blue nitrile
Taille
L
Stérilité
non-sterile
Caractéristiques
CE compliant
disposable
latex free
powder-free
Conditionnement
pack of 50 ea (boxes of 50)
Fabricant/nom de marque
KCL 743
L × épaisseur
270 mm × 0.2 mm
Description générale
Dermatril P is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril P disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril P disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
Caractéristiques et avantages
  • Anti-static
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Roughened fingertips for better grip
  • Rolled cuff edge for easier donning
  • CE compliant
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 61340-5-1, ASTM 1671:2001
Informations légales
Dermatril is a trademark of KCL GmbH
Les clients ont également consulté
Slide 1 of 1