Principaux documents
Z653330
Clips for Quickfit® flat-flange reaction flask lids
For 500mL
Synonyme(s) :
JC75F
Z653330-1EA
47.50 CHF
47.50 CHF
Check Cart for Availability
Changer de vue
Z653330-1EA
47.50 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
47.50 CHF
Check Cart for Availability
Matériaux
stainless steel
Étiquette/Marqueur
D
Stérilité
non-sterile
Conditionnement
pack of 1
Fabricant/nom de marque
Quickfit JC75F
Disponibilité
available only in EU
Alésage du collier
75 mm
Description générale
Designed for securing flat flange joints, these clips have a hinged ′spring wire′ design that tightens using a wing nut.
Informations légales
Quickfit is a registered trademark of SciLabware Limited