Principaux documents
Z549746
Sigma-Aldrich
Stopcock Adapter
straight glass stopcock, joint: ST/NS 34/45
1 EA
161.00 CHF
161.00 CHF
Check Cart for Availability
Changer de vue
1 EA
161.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
161.00 CHF
Check Cart for Availability
Matériaux
straight glass stopcock
Joint
joint: ST/NS34/45
Catégories apparentées
Description générale
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.