Principaux documents
Z531480
Sigma-Aldrich
Aldrich® Graham condenser
Overall H 600 mm, joint: ST/NS 24/40
1 EA
474.00 CHF
474.00 CHF
Check Cart for Availability
Changer de vue
1 EA
474.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
474.00 CHF
Check Cart for Availability
Joint
joint: ST/NS24/40
Disponibilité
not available in EU
H totale
600 mm
Description générale
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC