Principaux documents
Z530964
Sigma-Aldrich
Aldrich® Liebig condenser
Overall H 200 mm, joint: ST/NS 24/40
1 EA
144.00 CHF
144.00 CHF
Check Cart for Availability
Changer de vue
1 EA
144.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
144.00 CHF
Check Cart for Availability
Joint
joint: ST/NS24/40
H totale
200 mm
Catégories apparentées
Description générale
Condensers have a standard taper female joint and a lower male drip tip-joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC