Principaux documents
Z530387
Sigma-Aldrich
Aldrich® short-path distillation head
Joints: ST/NS 14/20 (2), thermometer joint: ST/NS 10/18
1 EA
94.00 CHF
94.00 CHF
Check Cart for Availability
Changer de vue
1 EA
94.00 CHF
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
94.00 CHF
Check Cart for Availability
Joint
thermometer joint: ST/NS10/18
Joints: ST/NS14/20 (2)
Profondeur d'immersion
50 mm
Catégories apparentées
Description générale
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC