Principaux documents
Z529540
Coors porcelain pestle
for Z529516
1 EA
160.00 CHF
160.00 CHF
Check Cart for Availability
Changer de vue
1 EA
160.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.35
160.00 CHF
Check Cart for Availability
Matériaux
porcelain
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Coorstek 66332
L du pilon
280 mm
Description générale
Oversized pestle glazed to grinding surface. Order mortar separately.
Application
The Coors porcelain pestle is suitable to use for crushing, grinding, and mixing.
Informations légales
Coors is a trademark of Coorstek, Inc.