Principaux documents
Z529052
Sigma-Aldrich
Aldrich® can/drum adapters
Bottle-top adapter, GL 45
1 EA
230.00 CHF
230.00 CHF
Check Cart for Availability
Changer de vue
1 EA
230.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.01
230.00 CHF
Check Cart for Availability
Catégories apparentées
Description générale
Chemically inert PBT adapters permit the attachment of pumps and bottle-top dispensers to Aldrich 5 gal cans or drums.
GL 45 to 2in. NPT thread. CERAMUS® to Aldrich 5 gallon container.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Ceramus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.