Principaux documents
Z516872
Sigma-Aldrich
Aldrich® two-neck round-bottom flask
capacity 250 mL, Joints: ST/NS 24/40 (2)
Synonyme(s) :
bottom flask, flask round, round flask
1 EA
108.00 CHF
108.00 CHF
Check Cart for Availability
Changer de vue
1 EA
108.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
108.00 CHF
Check Cart for Availability
Matériaux
borosilicate glass
round bottom flask
two-neck
Joint
Joints: ST/NS24/40 (2)
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
250 mL
Description générale
Angled ST joints. Z516929 and Z516953 have 24/40 side joints.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC