Principaux documents
Z503878
Scienceware® trays
w/o faucet, inside dimensions L × W × H 55 cm × 65 cm × 10.2 cm
1 EA
174.00 CHF
174.00 CHF
Check Cart for Availability
Changer de vue
1 EA
174.00 CHF
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.51
174.00 CHF
Check Cart for Availability
Matériaux
low-density polyethylene container
Fabricant/nom de marque
Bel-Art
Description générale
Molded LDPE can withstand temperatures up to 77°C (171°F). The PE faucet has a 12.7 mm bore.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP
Les clients ont également consulté
Slide 1 of 1