Principaux documents
Z415618
Sigma-Aldrich
Aldrich® side-arm distilling head
joint: ST/NS 24/40, thermometer joint: ST/NS 10/30
1 EA
191.00 CHF
191.00 CHF
Check Cart for Availability
Changer de vue
1 EA
191.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
191.00 CHF
Check Cart for Availability
Joint
thermometer joint: ST/NS10/30
joint: ST/NS24/40
Longueur du bras latéral
225 mm
Catégories apparentées
Description générale
A 50mm immersion length thermometer is recommended for the 14/20 joint, and 75mm immersion length thermometers are recommended for the 24/40 and 29/32 joints.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC