Principaux documents
Z312231
Gerhardt® STL56 warm air glassware dryers
AC/DC input 240 V AC, European 2-pin plug
About This Item
Code UNSPSC :
12352202
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Qualité
CE Compliant
Caractéristiques
Power: 900 W heater
Fabricant/nom de marque
Gerhardt
Disponibilité
not available in USA and Canada
Larg. × P × H
380 mm × 380 mm × 1150 mm
Description générale
Blows warm air up tubular supports for rapid drying of laboratory glassware. 56 white plastic-coated lateral drying tubes (24, 16 mm diam. 32 tubes, 11 mm diam.) on rotating central column. Electric heater and fan blows filtered hot or cold air up column and out of tube. Maximum temperature in glass vessel on dryer: approx 50 °C.
Heater top has raised lip to act as condensation/drip tray with drain pipe at rear. Front panel has power and heating indicator lamps, power switch, toggle switch for hot or cold air, and timer.
Supplied with Euro 2-pin plug.
Informations légales
Gerhardt is a registered trademark of Gerhardt