Principaux documents
Z274259
Sigma-Aldrich
Aldrich® tube fitting kit
for tubing i.d., 1/4 in.
1 KIT
216.00 CHF
216.00 CHF
Check Cart for Availability
Changer de vue
1 KIT
216.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
216.00 CHF
Check Cart for Availability
Caractéristiques
for tubing i.d. 1/4 in.
Description générale
Clear, natural Nylon-66 fittings for making a variety of connections with flexible tubing. Fittings are supplied in a compartmentalized PP storage box.
Contains 90 fittings that include: tubing hose barbs, connectors, tees, elbows, plugs, and locking sleeves.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC