Principaux documents
Z268763
Scienceware® Safe-Lab PTFE stirrer bearing
Replacement inserts
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.72
Matériaux
Viton® O-ring
Conditionnement
pkg of 2 ea
Fabricant/nom de marque
Bel-Art
Description générale
Stopper-Loc® nut allows stirrer bearing to be locked to outer ground-glass joint. Complete with Stopper-Loc nut and two PTFE inserts. For 10mm diam. polished glass or PTFE stirring shafts.
Informations légales
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Stopper-Loc is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company