Principaux documents
Z261491
Luer-to-Tubing Connector
bulkhead panel mounted Micro-Mate® female Luer to hose end for 1/16 in. to 3/32 in. I.D. tubing, nickel plated
Synonyme(s) :
environmental, veterinary
1 EA
56.30 CHF
56.30 CHF
Date d'expédition estimée le29 mai 2025
Changer de vue
1 EA
56.30 CHF
About This Item
Code UNSPSC :
40183110
Nomenclature NACRES :
NB.03
56.30 CHF
Date d'expédition estimée le29 mai 2025
Matériaux
nickel plated
Fabricant/nom de marque
Popper 6421
Description générale
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating, easy to clean and resistant to corrosion.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Informations légales
Micro-Mate is a registered trademark of Cadence Science