Principaux documents
Z244015
Collecteur ACE avec robinets d'arrêt filetés
positions, 3
1 EA
1 010.00 CHF
1 010.00 CHF
Check Cart for Availability
Changer de vue
1 EA
1 010.00 CHF
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.79
1 010.00 CHF
Check Cart for Availability
Caractéristiques
positions 3
Fabricant/nom de marque
Ace Glass 872950
Catégories apparentées
Description générale
Greaseless vacuum/gas manifold with 0 to 4mm high-vacuum PTFE, three-way stopcocks for takeoffs. One end of each manifold tube has a #7 Ace-Thred for easy cleaning. Ends are supplied with nylon stopper/plugs with FETFE® O-rings. Port connections are 8mm O.D. Vacuum and gas connections are 1/2in. o.d.
Informations légales
FETFE is a registered trademark of Ace Glass, Inc.