Principaux documents
Z215406
Rigid PTFE sleeve
joint: ST/NS 10/30
Synonyme(s) :
ptfe sleeves
1 EA
10.80 CHF
10.80 CHF
Check Cart for Availability
Changer de vue
1 EA
10.80 CHF
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
10.80 CHF
Check Cart for Availability
Joint
joint: ST/NS10/30
Fabricant/nom de marque
Cowie
Description générale
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.