Principaux documents
Z203696
Tube de seringue en PTFE
gauge 16, L 36 in.
1 EA
46.80 CHF
46.80 CHF
Check Cart for Availability
Changer de vue
1 EA
46.80 CHF
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.03
46.80 CHF
Check Cart for Availability
Longueur
36 in.
Calibre
16
Catégories apparentées
Description générale
PTFE syringe tubing with Kel-F Luer hub: A septum to be pierced with these PTFE needles must be punctured with a hollow SS needle. The PTFE needle is then threaded through. After threading, the SS needle is withdrawn, allowing the septum to seal around the PTFE needle.
For 20 ga PTFE use Z126756 (14 ga SS); 18 ga PTFE use Z126764 (13 ga SS); 16 ga PTFE use Z126772 (12 ga SS).
Autres remarques
PTFE tubing maximum temperature is 260 °C.