Principaux documents
Z178098
Scienceware® heavy-duty test-tube rack
Holds 32 x 16-20 mm tubes
1 EA
53.40 CHF
53.40 CHF
Check Cart for Availability
Changer de vue
1 EA
53.40 CHF
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.35
53.40 CHF
Check Cart for Availability
Matériaux
polypropylene
Caractéristiques
Holds 32 x 16-20 mm tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18857-0000
L × l × H
213 mm × 110 mm × 127 mm
Description générale
Natural PP, stackable. Steam autoclavable to 250°F (121°C). Rows lettered and numbered for easy identification. Base plate has rounded wells with drain holes; end plates with hand grips.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP