Principaux documents
Z172952
Sigma-Aldrich
Aldrich® pour-out adapter
male joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
male joint: ST/NS14/20
Description générale
For pouring from flasks or funnels without pouring over ground joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC