Principaux documents
Z156531
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 65/40
1 EA
150.00 CHF
150.00 CHF
Check Cart for Availability
Changer de vue
1 EA
150.00 CHF
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
150.00 CHF
Check Cart for Availability
Matériaux
stainless steel
Joint
joint: SJ65/40
Description générale
For use with Z13,730-8 and Z13,731-6
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.