Principaux documents
Z150681
Sigma-Aldrich
Window for Beckman® Multi-Cell Mount
NaCl, Drilled
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Matériaux
NaCl
rectangular
Description
Polished
Technique(s)
IR spectroscopy: suitable
L × l × H
29.5 mm × 14.5 mm × 4 mm
Informations légales
Beckman is a registered trademark of Beckman Coulter, Inc.
Multi-Cell Mount is a trademark of Beckman Coulter, Inc.