Principaux documents
Z146552
Ace Rotary-Evaporator/Freeze-Drying Flask Adapter Head W/ O-ring
joint: ST/NS 29/42
1 EA
241.00 CHF
241.00 CHF
Date d'expédition estimée le16 avril 2025
Changer de vue
1 EA
241.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
241.00 CHF
Date d'expédition estimée le16 avril 2025
Joint
joint: ST/NS29/42
Description
with FETFE®
O-ring.
Fabricant/nom de marque
Ace Glass 703023
Description générale
Flange with two ground flats opposite each other allows nylon retainers on adapter head to pass over. O-ring in head and flask grooves make a vacuum-tight seal when head is turned 90° in either direction. No grease, springs, or clamp necessary. Flasks sold separately.
Informations légales
FETFE is a registered trademark of Ace Glass, Inc.