Principaux documents
Z112488
Sigma-Aldrich
Magnetic holder
About This Item
Code UNSPSC :
23151816
Nomenclature NACRES :
SB.32
Technique(s)
IR spectroscopy: suitable
Description générale
  • Holds standard 13 mm KBr pellets and smaller pellets with paper inserts
  • Holds crystal windows listed (requires magnetic strips)