Principaux documents
Z107182
Sigma-Aldrich
Base plates
Unthreaded
1 EA
24.00 CHF
10 EA
203.00 CHF
24.00 CHF
Check Cart for Availability
Changer de vue
1 EA
24.00 CHF
10 EA
203.00 CHF
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
24.00 CHF
Check Cart for Availability
Description générale
Fits ½ in. lattice rods. Flat base.
These base plates are of a non-ferrous alloy, two inches in diameter, 16 mm high, with set screw for fastening lattice frame rods. Provided without mounting screws. Plate has four holes for attaching to bench, floor or wall.