Principaux documents
Z105082
Sigma-Aldrich
Spinbar® assortment
octagonal PTFE-coated, pack of 12 ea
12 EA
120.00 CHF
120.00 CHF
Check Cart for Availability
Changer de vue
12 EA
120.00 CHF
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
120.00 CHF
Check Cart for Availability
Matériaux
octagonal PTFE-coated
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art F37182-0000
Description générale
2 of each size. 1/2 × 5/16 in., 11/2 × 3/8 in., 5/8 × 5/16 in., 2 × 3/8 in., 1 × 3/8 in., 21/2 × 3/8 in.
Informations légales
Spinbar is a registered trademark of SP Scienceware, a division of SP