Principaux documents
WHA77001101
Whatman® UNIFILTER® 384 well filter plates
clear polystyrene, non-sterile, dye-terminator, 50 ea
Synonyme(s) :
384 well plates, Z746363, micro plates, well plates
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.22
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Nom du produit
Whatman® UNIFILTER® plates 384, size 384 wells, Dye removal plate, well maximum volume 100 μL, GF/C, clear polystyrene, filter bottom with long drip director
Matériaux
conical bottom natural wells
natural polypropylene
polystyrene membrane
Caractéristiques
GF/C, clear polystyrene, filter bottom with long drip director
Conditionnement
pkg of 50 ea
Fabricant/nom de marque
Whatman 7700-1101
Whatman Article No. 28420789 (US reference)
Taille
384 wells
Volume maximum des puits
100 μL
Volume de travail par puits
100 μL
Dimension de pores
0.45 μm
Type de liaison
non-treated surface
Description générale
Cytiva′s Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Polystyrene microplate for clean-up and cell capture
A high-throughput microplate can facilitate cell capture or dye removal and other sample clean-up processes requiring high recovery. Whatman UNIFILTER microplates are a good choice because of their long drip director and their ability to support gel filtration media.
Caractéristiques et avantages
  • Usable with gel filtration media, allowing high-throughput removal of dye blobs
  • Long drip director prevents well-to-well cross contamination
  • Rigid, clear polystyrene microplate can withstand centrifugation
  • Membrane media option of DNA binding polymer or glass microfiber
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva