Principaux documents
WHA2800432
Whatman® high performance cellulose extraction thimbles
single thickness, I.D. × H 43 mm × 123 mm
Synonyme(s) :
Z741973, cellulose filter, paper filter, whatman filter
About This Item
Code UNSPSC :
41104817
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Whatman 2800-432
Whatman Article No. 28418983 (US reference)
D.I. × H
43 mm × 123 mm
Catégories apparentées
Description générale
Cytiva′s Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.
Cellulose thimble options for Soxhlet extractor systems
Standard extraction thimbles have a wall thickness of 1–1.5 mm. Double-thickness thimbles have a wall thickness of approximately 2 mm for applications that require higher retention, increased wet or dry strength, or increased rigidity. Measurements can be matched to specific Soxhlet extractor systems.
Caractéristiques et avantages
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120°C (248°F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1–1.5 mm to support application need
  • Option for double thickness for added rigidity
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva