Principaux documents
TEM-Q350AR1.3-2NM
Sigma-Aldrich
QUANTIFOIL R 1.2/1.3 with Ultra-Thin Carbon
Gold Grids, size 300 mesh, pack of 50
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-Q350AR1.3-2NM n'est pas disponible actuellement. Contacter notre Service technique
Conditionnement
pack of 50
Taille
300 mesh
Description générale
QUANTIFOIL with circular holes is used in cryoelectron tomographic reconstruction. The roundness of the holesis advantageous with respect to the formation of an icelayer of constant thickness. Assessment of the image quality is easier when foil is incl
Questions
  1. What is the thickness of the coated carbon layer?
    1 answer
    1. The foil thickness of the coated carbon layer is ~12nm thick