Principaux documents
TEM-CFLF135-AU-25
Sigma-Aldrich
Carbon Film Supported Gold London Finder LF135 Grids
box of 25
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-CFLF135-AU-25 n'est pas disponible actuellement. Contacter notre Service technique
Conditionnement
box of 25
Description générale
A 5-6 nm thin film of pure carbon supported London finder (LF135) grids. Gold grid. Carbon film show better electrical and heat conduction in the electron beam. London finder grids are locator grids with reference/index on the grid.