Principaux documents
TEM-CF300CU50
Sigma-Aldrich
Carbon Film Supported Copper Grid
Standard Thickness, size 300 mesh, box of 50 ×
Synonyme(s) :
Carbon Film 300 Mesh, Copper
About This Item
Code UNSPSC :
41111761
Matériaux
carbon film
copper grid
Conditionnement
box of 50 ×
film size
5-6 nm
Taille
300 mesh
Description générale
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.
Remplacé(e)(s) par