Principaux documents
SYNV130711
Synthware conical bottom reaction vial with holed compression cap
1 mL, joint: ST/NS 7/10
Synonyme(s) :
Synthware V130711, reaction tube
1 EA
22.20 CHF
22.20 CHF
Check Cart for Availability
Changer de vue
1 EA
22.20 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
22.20 CHF
Check Cart for Availability
Joint
joint: ST/NS7/10
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware V130711
Capacité
1 mL
Catégories apparentées
Description générale
Supplied with one holed compression cap, one Viton® O-ring and one PTFE -faced silicone rubber septum. For replacement caps, see RC112; For Viton® O-rings, see R014; for septa, see R1420.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company