Principaux documents
SYNS342565
Synthware high vacuum oblique glass stopcock
bore size 8 mm, plug size 25/65
Synonyme(s) :
Synthware S342565
1 EA
137.00 CHF
137.00 CHF
Check Cart for Availability
Changer de vue
1 EA
137.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
137.00 CHF
Check Cart for Availability
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S342565
Taille de l'alésage
8 mm
Description générale
Precision ground with corresponding numbers on the barrel and plug. Lower vacuum chamber is evacuated via a separate 2 mm hole in the plug which is not in the line of flow.
Informations légales
Synthware is a trademark of Kemtech America Inc.