Principaux documents
SYNM131519
Synthware #15 o-ring joint to inner joint adapter
joint: ST/NS 19/22
Synonyme(s) :
Synthware M131519, glass joint adapter
1 EA
34.00 CHF
34.00 CHF
Check Cart for Availability
Changer de vue
1 EA
34.00 CHF
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
34.00 CHF
Check Cart for Availability
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware M131519
Catégories apparentées
Description générale
Synthware adapter with #15 O-Ring and an inner joint (19/22) to inner ground glass.
Informations légales
Synthware is a trademark of Kemtech America Inc.