Principaux documents
SYNF641125
Synthware pressure equalizing funnel with high vacuum valve
125 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonyme(s) :
Synthware F671425, dropping funnel
1 EA
177.00 CHF
177.00 CHF
Check Cart for Availability
Changer de vue
1 EA
177.00 CHF
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
177.00 CHF
Check Cart for Availability
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F641125
Capacité
125 mL
125 mL
Catégories apparentées
Description générale
With PTFE extended tip valve for semi-needle valve adjustment.
Informations légales
Synthware is a trademark of Kemtech America Inc.