Principaux documents
SYNF591025
Synthware three-neck round bottom flask
25 mL, joint: ST/NS 14/10
Synonyme(s) :
Synthware F5910125, 3-neck flask
1 EA
70.70 CHF
70.70 CHF
Check Cart for Availability
Changer de vue
1 EA
70.70 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
70.70 CHF
Check Cart for Availability
Matériaux
borosilicate glass flask
round bottom
three-neck
Joint
joint: ST/NS14/10
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F591025
Capacité
25 mL
25 mL
Description générale
Microscale, round bottom flask with three 14/10 outer ground joints externally threaded. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company