Principaux documents
SYNF561050
Synthware reaction tube with high vacuum valve
50 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonyme(s) :
Synthware F561050, Schlenk flask
1 EA
102.00 CHF
102.00 CHF
Check Cart for Availability
Changer de vue
1 EA
102.00 CHF
About This Item
Code UNSPSC :
41121700
Nomenclature NACRES :
NB.51
102.00 CHF
Check Cart for Availability
Matériaux
colorless tube
glass tube
Joint
joint: ST/NS14/20
Stérilité
non-sterile
Caractéristiques
graduations: no
Push fit
Conditionnement
pack of 1 ea
pkg of 1 ea
Fabricant/nom de marque
Kemtech F561050
Capacité
50 mL
50 mL
Taille de la vanne
0-4 mm
Description générale
Supplied with high vacuum valve with PTFE o-ring at tip and PTFE wiper (“umbrella”) to protect Viton® O-ring on shaft.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company