Principaux documents
SYNF484258
Synthware solvent transfer/storage flask
250 mL, joint: ST/NS 24/40, valve size 0-8 mm
Synonyme(s) :
Synthware F484258, Straus flask, receiver flask
1 EA
118.00 CHF
118.00 CHF
Check Cart for Availability
Changer de vue
1 EA
118.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
118.00 CHF
Check Cart for Availability
Matériaux
borosilicate glass flask
round bottom
two-neck
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F484258
Capacité
250 mL
250 mL
Taille de la vanne
0-8 mm
Description générale
Synthware solvent transfer/storage flask features a center neck joint sealed off from direct access to flask. A syringe or cannula is used to recover the solvent through the valve.
Informations légales
Synthware is a trademark of Kemtech America Inc.