Principaux documents
SYNF434100
Synthware three neck round bottom flask with angled side necks
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Synonyme(s) :
Synthware F434100, 3-neck round flask
1 EA
80.20 CHF
80.20 CHF
Check Cart for Availability
Changer de vue
1 EA
80.20 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
80.20 CHF
Check Cart for Availability
Matériaux
borosilicate glass flask
round bottom
three-neck (angled side necks)
Joint
center joint: ST/NS24/40
side joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F434100
Capacité
100 mL
100 mL
Catégories apparentées
Description générale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.