Principaux documents
SYNF431500
Synthware three neck round bottom flask with angled side necks
500 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Synonyme(s) :
Synthware F431500, 3-neck round flask
1 EA
83.00 CHF
83.00 CHF
Check Cart for Availability
Changer de vue
1 EA
83.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
83.00 CHF
Check Cart for Availability
Matériaux
borosilicate glass flask
round bottom
three-neck
Joint
center joint: ST/NS14/20
side joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F431500
Capacité
500 mL
500 mL
Description générale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.