Principaux documents
SYNF304250
Synthware single-neck round-bottom flask
250 mL, joint: ST/NS 24/40
Synonyme(s) :
Synthware F304250, glass round flask
1 EA
31.60 CHF
31.60 CHF
Check Cart for Availability
Changer de vue
1 EA
31.60 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
31.60 CHF
Check Cart for Availability
Matériaux
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F304250
Capacité
250 mL
250 mL
Description générale
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.