Principaux documents
SYNF150050
Synthware micro filter flask
capacity 50 mL, stopper size #3
Synonyme(s) :
Synthware F150050
1 EA
23.50 CHF
23.50 CHF
Check Cart for Availability
Changer de vue
1 EA
23.50 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
23.50 CHF
Check Cart for Availability
Matériaux
flat bottom flask
glass flask
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F150050
Capacité
50 mL
Description générale
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Informations légales
Synthware is a trademark of Kemtech America Inc.