Principaux documents
SYND191420
Synthware distilling receiver with hose connection
joint: ST/NS 14/20
Synonyme(s) :
Synthware D191420, glass distilling receiver
1 EA
130.00 CHF
130.00 CHF
Check Cart for Availability
Changer de vue
1 EA
130.00 CHF
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.51
130.00 CHF
Check Cart for Availability
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware D191420
Catégories apparentées
Description générale
Cow-type design having three receivers spaced 40o apart. 105o angle between top outer joint and lower inner joints.
Informations légales
Synthware is a trademark of Kemtech America Inc.