Principaux documents
SYNC171200
Synthware water jacketed distillation column with indents to support packing material
joint: ST/NS 14/20, jacket H 200 mm
Synonyme(s) :
Synthware C171200, jacketed glass distilling column
1 EA
66.00 CHF
66.00 CHF
Check Cart for Availability
Changer de vue
1 EA
66.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
66.00 CHF
Check Cart for Availability
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C171200
H de l'enveloppe
200 mm
Catégories apparentées
Description générale
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Informations légales
Synthware is a trademark of Kemtech America Inc.