Principaux documents
SYNA902440
Synthware Claisen distillation adapter with compression cap thermometer port
joint: ST/NS 24/40
Synonyme(s) :
Synthware A902440, Glass Distillation Adapter Claisen Thermometer
1 EA
102.00 CHF
102.00 CHF
Check Cart for Availability
Changer de vue
1 EA
102.00 CHF
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
102.00 CHF
Check Cart for Availability
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A902440
Catégories apparentées
Description générale
With threaded thermometer port, supplied complete with Nylon cap and Viton® O-ring to accommodate 6-7.5 mm O.D. thermometer.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company