Principaux documents
SLW1523/18
Pyrex® Media-Lab Bottles, wide mouth, with polypropylene cap and pouring ring, with printed trace code
capacity 10000 mL
Synonyme(s) :
Pyrex, SciLabware 1523/18
1 EA
396.00 CHF
396.00 CHF
Check Cart for Availability
Changer de vue
1 EA
396.00 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
396.00 CHF
Check Cart for Availability
Matériaux
borosilicate glass bottle
polypropylene cap
round bottle
white graduation
wide-mouth
Joint
Joints threaded neck (GLS80)
Stérilité
non-sterile
Caractéristiques
graduations
with cap
with pouring ring
with printed trace code
Conditionnement
pack of 1 ea
Fabricant/nom de marque
SciLabware 1523/18
Hauteur
389 mm
Diam. du corps
227 mm
Capacité
10000 mL
Description générale
  • Bottles complete with polypropylene screwcap and drip-free pouring ring
  • Manufactured from Pyrex borosilicate glass with proven temperature and chemical resistance qualities
  • GLS 80 screwthread provides increased access compared to GL 45 bottles aiding filling and pouring
  • Ideal for use with viscous liquids, pastes, powders, granules and larger solids
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White enamel graduations and marking spot
  • Autoclavable
  • Screwcaps and pouring rings can withstand temperatures up to 140 deg. C
Informations légales
PYREX is a registered trademark of Corning, Inc.